Drukuj

Anti-LRRC19 Rabbit Polyclonal Antibody

Dostawca: Abgent
ABGEAP19872BEA 1750 PLN
ABGEAP19872B
Anti-LRRC19 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: LRRC19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR