Chcesz zalogować się jeszcze raz?
Anti-ELAC1 Rabbit Polyclonal Antibody
Dostawca: Abgent
Type: Primary
Antigen: ELAC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
ZAMÓW
Rozwiń / Zwiń
Ilość Numer katalogowy VWR Ilość
400 µl ABGEAP17571A
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF
SPECYFIKACJE
Antigen symbol ELAC1
Antigen name elaC ribonuclease Z 1
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Flow cytometry Yes
Antigen synonyms D29
Amino acid number 73 to 101
Gene ID 55520
Epitope N-terminal
Stężenie 0.55
Immunogen This ELAC1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 73-101 amino acids from the N-terminal region of human ELAC1.
Molecular weight 40.019 kDa
Purification Protein A & Peptide affinity purification
Sequence KQPIEIYGPVGLRDC
Storage buffer pH 7,4
Temperatura przechowywania Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at – 20 °C in small aliquots to prevent freeze-thaw cycles
Shipping temperature 4 °C
Wprowadź szczegóły
Select the article for which you are looking for Certificate
Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość