Chcesz zalogować się jeszcze raz?
Anti-PEAK1 Rabbit Polyclonal Antibody
Dostawca: Abgent
Type: Primary
Antigen: PEAK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
ZAMÓW
Rozwiń / Zwiń
Ilość Numer katalogowy VWR Ilość
400 µl ABGEAP18239A
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF
SPECYFIKACJE
Antigen symbol PEAK1
Antigen name pseudopodium-enriched atypical kinase 1
Reactivity Human, Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Western blot Yes
ELISA Yes
Flow cytometry Yes
Antigen synonyms SGK269
Amino acid number 673 to 699
Gene ID 79834
Epitope N-terminal
Stężenie 0.65
Immunogen This SGK269 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 673-699 amino acids from the N-terminal region of human SGK269.
Molecular weight 193.106 kDa
Purification Protein A & Peptide affinity purification
Sequence TKGFSNSTEHKRC
Storage buffer pH 7,4
Temperatura przechowywania Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at – 20 °C in small aliquots to prevent freeze-thaw cycles
Shipping temperature 4 °C
Wprowadź szczegóły
Select the article for which you are looking for Certificate
Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość