Drukuj
Anti-ZNF703 Rabbit Polyclonal Antibody
Dostawca: Abgent
ABGEAP18351CEA 1750 PLN
ABGEAP18351C
Anti-ZNF703 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: ZNF703
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human