Drukuj

Anti-NAA60 Rabbit Polyclonal Antibody

Dostawca: Abgent
ABGEAP18377CEA 1750 PLN
ABGEAP18377C
Anti-NAA60 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: NAA60
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR