Drukuj
Anti-ZNF696 Rabbit Polyclonal Antibody
Dostawca: Abgent
ABGEAP19572AEA 1750 PLN
ABGEAP19572A
Anti-ZNF696 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: ZNF696
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human