Chcesz zalogować się jeszcze raz?
Anti-UBE2Z Rabbit Polyclonal Antibody
Dostawca: Abgent
Type: Primary
Antigen: UBE2Z
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
ZAMÓW
Rozwiń / Zwiń
Ilość Numer katalogowy VWR Ilość
400 µl ABGEAP17226B
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF
SPECYFIKACJE
Antigen symbol UBE2Z
Antigen name Ubiquitin-conjugating enzyme E2Z
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Antigen synonyms USE1, HOYS7
Amino acid number 284 to 310
Gene ID 65264
Epitope C-terminal
Stężenie 0.72
Immunogen This UBE2Z antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 284-310 amino acids from the C-terminal region of human UBE2Z.
Molecular weight 38.21 kDa
Purification Protein A & Peptide affinity purification
Sequence DPFGEKRGHFDYC
Storage buffer pH 7,4
Temperatura przechowywania Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at – 20 °C in small aliquots to prevent freeze-thaw cycles
Shipping temperature 4 °C
Wprowadź szczegóły
Select the article for which you are looking for Certificate
Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość