Drukuj
Anti-NT5DC2 Rabbit Polyclonal Antibody
Dostawca: Abgent
ABGEAP18155CEA 1750 PLN
ABGEAP18155C
Anti-NT5DC2 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: NT5DC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human