Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-RMND5A Rabbit Polyclonal Antibody

Dostawca: Abgent
Type: Primary
Antigen: RMND5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse




 

ZAMÓW

Rozwiń / Zwiń
Ilość Numer katalogowy VWR Ilość
400 µl ABGEAP17828B
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol RMND5A
Antigen name required for meiotic nuclear division 5 homolog A
Reactivity Mouse, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Antigen synonyms p44CTLH, GID2A, RMD5, CTLH, GID2
Amino acid number 300 to 327
Gene ID 64795
Epitope C-terminal
Stężenie 0.6
Immunogen This RMND5A antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 300-327 amino acids from the C-terminal region of human RMND5A.
Molecular weight 43.993 kDa
Purification Protein A & Peptide affinity purification
Sequence KDELPIEVDLGKKC
Storage buffer pH 7,4
Temperatura przechowywania Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at – 20 °C in small aliquots to prevent freeze-thaw cycles
Shipping temperature 4 °C
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość