Drukuj

Yttrium(III) carbonate hydrate ≥99.9% (REO, rare earth oxide basis)

Dostawca: Thermo Scientific

40651.18EA 850 PLN
40651.18 40651.30
Yttrium(III) carbonate hydrate ≥99.9% (REO, rare earth oxide basis)
Yttrium(III) carbonate hydrate
Formuła: Y₂(CO₃)₃·xH₂O
MW: 357,84 g/mol (anhy)
Temperatura przechowywania: Temperatura otoczenia
Numer MDL: MFCD00149943
Numer CAS: 38245-39-5
Numer WE (dawny EINECS): 209-119-4

Zobacz ceny


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR