Drukuj

Sodium potassium (K₂Na)-silica gel 35 - 40% alkali metal in silica gel Stage I

Niebezpieczeństwo

44890.14EA 1520 PLN
44890.14 44890.06
Sodium potassium (K₂Na)-silica gel 35 - 40% alkali metal in silica gel Stage I
Sodium potassium (K₂Na)-silica gel

Applications: Reductively produce radical anions and dianions, dechlorinate alkyl and aryl halides and desulfurize compounds

Gęstość: 0,7
Temperatura przechowywania: Temperatura otoczenia
Numer MDL: MFCD08705348
Jednostka: 2813
ADR: 4.3,II

Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR