Drukuj
Melanocyte-stimulating hormone-release inhibiting factor
Dostawca: Thermo Fisher Scientific
J60045.03EA 1420 PLN
J60045.03 J60045.MD
Melanocyte-stimulating hormone-release inhibiting factor
Białka
CAS No.: 2002-44-0
Molecular Formula: C13H24N4O3
Formula Weight: 284.36
Storage Temperature: −30 to −10 °C
Physical Form: Lyophilized powder
Appearance: White to off-white
MDL No.: MFCD00037866
Applications: Blocks the release of alpha-melanocyte stimulating hormone, increases brain dopamine levels and antagonizes physiological and behavioral opioid effects in vivo