Drukuj

Human angiotensin 1

Dostawca: Thermo Fisher Scientific
J62102.MBEA 1780 PLN
J62102.MB
Human angiotensin 1
Białka
CAS No.: 484-42-4
Molecular Formula: C62H89N17O14
Formula Weight: 1296,50
Storage Temperature: −30 to −10 °C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00133091
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR