Drukuj

Oxytocin

Dostawca: Thermo Fisher Scientific
J63421.MBEA 6350 PLN
J63421.MB
Oxytocin
Białka
Peptide hormone that acts predominantly as a neurotransmitter; stimulates uterine contraction and lactation.

  • EINECS: 200-048-4
  • GHS symbols and signal word: GHS06, GHS08 and Danger
  • Hazard class and packing group: 6.1, II
  • UN number: 2811
  • Merck index number: 14,6979
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR