Drukuj

G3335

Dostawca: Thermo Fisher Scientific
J64528.MFEA 982 PLN
J64528.MF
G3335
Białka
  • Light sensitive

CAS No.: 36099-95-3
Molecular Formula: C16H19N3O5
Formula Weight: 333,34
Storage Temperature: 2 to 8 °C
Physical Form: Solid
Appearance: White
MDL No.: MFCD00037964

Applications: A cell-permeable dipeptide that acts as a selective and reversible peroxisome proliferator-activated receptor antagonist
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR