/
Poland
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Nitrite standard solution
HACH2340249
:
Hach
:
Nitrite standard solution
HACH2340249
:
Hach
:
399 zł
1 * 500 mL
-
+
Concentration:
250 mg/l as N
Pk:
500 mL
Description:
Nitrite standard solution, stock, 500 ml
For accuracy checks in nitrite determinations.