Order Entry
Multi-channel pipettes, mechanical, variable volume, Tacta
 :  Sartorius
undefined
Multi-channel pipettes, mechanical, variable volume, Tacta
 :  Sartorius
  • Catalog No:
  • 613-5488
  • 613-5487
  • 613-5486
  • 613-5491
  • 613-5490
  • 613-5489
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
8-channel
  •  
    Volume
    Increment
    Accuracy
    Imprecision
    Colour code
    With Safe-Cone filter(s)
  • 613-5486
    LH-729120
    1 - 10 µl
    0,01
    1,50 - 4,00 %
    1,00 - 4,00 %
    Grey
    -
    1 * 1 SZT
    1 * 1 SZT - 4 920 zł
    4 920 zł
    1 * 1 SZT
       4 920 zł
     : LH-729120
     : BHITLH-729120
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-5487
    LH-729130
    10 - 100 µl
    0,10
    0,70 - 3,00 %
    0,25 - 1,50 %
    Yellow
    613-4071, 613-0694*
    1 * 1 SZT
    1 * 1 SZT - 4 920 zł
    4 920 zł
    1 * 1 SZT
       4 920 zł
     : LH-729130
     : BHITLH-729130
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-5488
    LH-729140
    30 - 300 µl
    0,20
    0,60 - 2,00 %
    0,25 - 1,00 %
    Orange
    613-4070, 613-0693
    1 * 1 SZT
    1 * 1 SZT - 4 920 zł
    4 920 zł
    1 * 1 SZT
       4 920 zł
     : LH-729140
     : BHITLH-729140
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
12-Channel
  •  
    Volume
    Increment
    Accuracy
    Imprecision
    Colour code
    With Safe-Cone filter(s)
  • 613-5489
    LH-729220
    1 - 10 µl
    0,01
    1,50 - 4,00 %
    1,00 - 4,00 %
    Grey
    -
    1 * 1 SZT
    1 * 1 SZT - 5 560 zł
    5 560 zł
    1 * 1 SZT
       5 560 zł
     : LH-729220
     : BHITLH-729220
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-5490
    LH-729230
    10 - 100 µl
    0,10
    0,70 - 3,00 %
    0,25 - 1,50 %
    Yellow
    613-4071, 613-0694*
    1 * 1 SZT
    1 * 1 SZT - 5 560 zł
    5 560 zł
    1 * 1 SZT
       5 560 zł
     : LH-729230
     : BHITLH-729230
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-5491
    LH-729240
    30 - 300 µl
    0,20
    0,60 - 2,00 %
    0,25 - 1,00 %
    Orange
    613-4070, 613-0693
    1 * 1 SZT
    1 * 1 SZT - 5 560 zł
    5 560 zł
    1 * 1 SZT
       5 560 zł
     : LH-729240
     : BHITLH-729240
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact