Skip to Header
Skip to Main Content
Skip to Footer
/
Poland
Products
Services
Digital Solutions
Applications
Support
About
0 zł
Order Entry
Home
Buffers
Assay Buffers
Gelatin veronal buffer with EDTA (GVBE), 5 mM barbital
Gelatin veronal buffer with EDTA (GVBE), 5 mM barbital
Gelatin veronal buffer with EDTA (GVBE), 5 mM barbital
:
Thermo Fisher Scientific
Gelatin veronal buffer with EDTA (GVBE), 5 mM barbital
:
Thermo Fisher Scientific
Liquid.
Thermo Scientific Acroseal 25 Years