Anaerobic systems, Genbag
BMER45532
- Description:GENbag, microaerophil
- Pk:20 Tests
Transparent bag, for incubation of single specimens. The cultures can be observed without opening the system.
- No additional material required
- No hydrogen release thereby making them completely user-safe
- High-performance: atmosphere required in less than one hour