Order Entry
Anaerobic systems, Genbag
  BMER45532
Anaerobic systems, Genbag
  BMER45532
  1 * 20 Tests
This product has been blocked by your organization. Please contact your purchasing department for more information
  • Description:
    GENbag, microaerophil
  • Pk:
    20 Tests
Transparent bag, for incubation of single specimens. The cultures can be observed without opening the system.
  • No additional material required
  • No hydrogen release thereby making them completely user-safe
  • High-performance: atmosphere required in less than one hour