Drukuj
Podłoła hodowlane komórek macierzystych, HyClone™ HyCell-STEM™
Dostawca: HyClone products (Cytiva)
HyClone™
HYCLSR30004.01EA 1180 PLN
HYCLSR30004.01 HYCLSR30003.01
Podłoła hodowlane komórek macierzystych, HyClone™ HyCell-STEM™
Pożywki do hodowli komórkowych
HyClone™ HyCell-STEM™ and HyCell-STEM-FF™ complete media have been developed for the culture of human embryonic stem cells (hESCs) and induced pluripotent stem (hiPS) cells. HyCell-STEM™ is intended for use with stem cells on feeder layer cultures while HyCell-STEM-FF™ is designed for feeder-free conditions. Both media have been shown to increase cell growth with fewer passages without compromising pluripotency.
  • Increase rate of expansion
  • Serum-free formulation
  • Maintain stemness
  • Improved post-thaw recovery
  • Sustain healthy stem cell morphology
Dodatkowe informacje: Kit includes 6X supplement and DMEM/F12 to create a complete media. Basic fibroblast growth factor (bFGF, 10 ng/ml) must be added to the completed media at time of use.