Wash buffer, Pierce™
These wash buffers are designed for use in dialysis, cross-linking, enzyme assays, ELISAs, immunohistochemistry, protein plate-coating, biotinylation applications.
- Ready to use without having to reconstitute with ultrapure water
- Eliminates the possibility of powder remaining in a packet
- 10X or 20X concentration eliminates time spent waiting for powder to dissolve
- Storage as concentrated stock minimizes bench space needed for large volume solutions
- pH valuePk
- Pierce™ borate buffer, 20XPierce™ borate buffer, 20XPIER28341283418.50,5 L1 * 0,5 L674 zł
- Pierce™ PBS Tween™ 20 buffer, 20XPierce™ PBS Tween™ 20 buffer, 20XPIER28352283527,50,5 L1 * 0,5 L663 zł
- Pierce™ TBS Tween™ 20 buffer, 20XPierce™ TBS Tween™ 20 buffer, 20XPIER28360283607,50,5 L1 * 0,5 L674 zł
- Pierce™ modified Dulbecco's PBS buffer, 20XPierce™ modified Dulbecco's PBS buffer, 20XPIER28344283447,40,5 L1 * 0,5 L758 zł
- Pierce™ modified Dulbecco's PBS Tween™ 20 buffer, 20XPierce™ modified Dulbecco's PBS Tween™ 20 buffer, 20XPIER28346283467,40,5 L1 * 0,5 L764 zł