This device is designed for the measurement of conductivity, resistivity, salinity and TDS in liquids − using a permanently connected electrode (measuring cell). It is suitable for mobile applications and for stationary operation in controlled electromagnetic environments (laboratories).
- Robust ABS case (IP 65)
- Min./Max./HOLD function
- Alarm for low battery and re-calibration
- Auto-Off function (adjustable: 1 - 120 min.)
- Clip allows belt attachment, benchtop use and wall-mounting
- Serial interface (3,5 mm jack) can be connected to the RS232 or USB interface of a PC via an electrically isolated interface converter (not included)
Dodatkowe informacje: Supplied with conductivity/temperature cell, one standard bottle, case, 9 V battery and manual.