Biotin NTA
Supplier: Biotium
Certificates
About this item
Biotinylated probes for detection of His-tagged proteins immobilised on nitrocellulose membranes.
- White solid soluble in water
- Removable from the histidine-tagged protein at pH 4,8
Biotin NTA, also known as BNTA (Biotin nitrilotriacetic acid, tripotassium salt) reversibly binds to histidine-tagged proteins, and can be detected with Biotium's CF™ Dye streptavidin or monoclonal anti-biotin fluorescent conjugates. Available with 6-atom aminohexanoyl (X) linker between the biotin and NTA to facilitate interaction with streptavidin.
Product Information
- Catalog #Supplier #PkAvailabilityPrice
- Biotin NTA or BNTA (Biotin nitrilotriacetic acid, tripotassium salt)Biotin NTA or BNTA (Biotin nitrilotriacetic acid, tripotassium salt)90074.900745 mg1 * 5 mg1 010 złSupplier Article Number: 90074Local Article Number: BTIU90074
- Biotin-X NTA (Biotin-X nitrilotriacetic acid, tripotassium salt)Biotin-X NTA (Biotin-X nitrilotriacetic acid, tripotassium salt)90056.900565 mg1 * 5 mg1 010 złSupplier Article Number: 90056Local Article Number: BTIU90056