/
Poland
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Bovine serum albumin (BSA), Fraction V
:
Thermo Fisher Scientific
Bovine serum albumin (BSA), Fraction V
:
Thermo Fisher Scientific
Light yellow to light beige Powder
Source:
Species:
Environmentally Preferable:
ACRO24040
Pack type
Pk
ACRO240401000
240401000
Glass bottle
100 g
1 * 100 g
1 * 100 g - 0 zł
0 zł
-
+
1 * 100 g
0 zł
-
+
ACRO240405000
240405000
Glass bottle
500 g
1 * 500 g
1 * 500 g - 6 810 zł
6 810 zł
-
+
1 * 500 g
6 810 zł
-
+