Drukuj
Paski mikropłytek immunologiczne, Nunc™ NucleoLink™
Dostawca: Thermo Fisher Scientific
Nunc™ Nunc™ NucleoLink™
732-2612EA 714 PLN
NUNC248259
Paski mikropłytek immunologiczne, Nunc™ NucleoLink™
NucleoLink™ strips are ideal for binding nucleotides (eg. DNA and RNA) and for hybridisation assays.
  • V-shaped wells with flat, readable bottoms and thin walls (0,35 mm)
  • Usable with any 'ELISA-like' detection and measurement method
  • Consists of 1×8 strips to be fitted into a 96-well format frame
  • Withstand temperatures from –20 to +120 °C
NucleoLink™ breakable modules are ideal for hybridisation assays due to the heat resistant resin. Each well locks into the frame and maintains the same height to provide accurate reading and efficient washing.
Sposób pakowania: Individually packed, 1440 per case (corresponds to 120 frames).