Drukuj
Szybki żelowy bufor separacyjny RNA, 10X
1B1373-500MLEA 381 PLN
VWRC1B1373-500ML
Szybki żelowy bufor separacyjny RNA, 10X
RNA Gel running buffer is typically used in denaturing formaldehyde RNA gels.
  • Faster gel results
  • Easy-to-use replacement for standard MOPS buffer
It is specially formulated to allow application of higher voltages during electrophoresis without causing overheating or gel distortion. RNA can be resolved twice as fast without adding any additional protocol steps or new equipment purchases.