Poland
Order Entry
Rubidium chloride 99.99% (trace metals basis)
Rubidium chloride 99.99% (trace metals basis)
  ACRO193920100
 :  
undefined
Rubidium chloride 99.99% (trace metals basis)
  ACRO193920100
 :  
 : 193920100
 : ACRO193920100
 
1 460 zł
   
  1 * 10 g

 

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

 

 

  • Appearance:
    White Powder and/or chunks
  • Titration Argentometric:
    28.9 to 30.1 % (Cl)
  • Total trace metal impurities:
    ≤100 ppm

 

 

 :
 :120.92 g/mol
 :1390 °C (760 mmHg)
 :718 °C
 :2,8 g/cm³ (20 °C)
 :Ambient
 :7791-11-9
 :RbCl
 :MFCD00011187