Order Entry
Temperature gradient plate for MFFT test
Temperature gradient plate for MFFT test
Supplier:  Industrial Physics
Certificates
undefined
Temperature gradient plate for MFFT test
Supplier:  Industrial Physics
Related Information
Product Information
  • Catalog #
    Supplier #
    Pk
    Availability
    Price
  • MFFT 10, temperature range −5...+80 °C, with a maximum gradient on surface of 20 °C
    MFFT 10, temperature range −5...+80 °C, with a maximum gradient on surface of 20 °C
    SIMEVF9600
    VF9600
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 146 830 zł
    146 830 zł
    1 * 1 SZT
       146 830 zł
    Supplier Article Number: VF9600
    Local Article Number: SIMEVF9600
    Specifications:
    Specifications
    Environmentally Preferable
  • MFFT 20- Temperature range of −30...+250 °C with a maximum gradient on surface of 100 °C
    MFFT 20- Temperature range of −30...+250 °C with a maximum gradient on surface of 100 °C
    SIMEVF9700
    VF9700
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 230 380 zł
    230 380 zł
    1 * 1 SZT
       230 380 zł
    Supplier Article Number: VF9700
    Local Article Number: SIMEVF9700
    Specifications:
    Specifications
    Environmentally Preferable
Frequently Bought Together