Drukuj
Wyszukano: Wzorce biologiczne
125  wyniki/ów wyszukiwania
SearchResultCount:"125"
Sort Results
Widok listy Widok uproszczony (nowość)
Oceń wyniki wyszukiwania
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
Numer katalogowy: (1535824.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
j.m.: 1 * 1 SZT
Numer katalogowy: (1048244.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
j.m.: 1 * 500 mg
Numer katalogowy: (1509019.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
j.m.: 1 * 1 g
Numer katalogowy: (1302258.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
j.m.: 1 * 1 g
Numer katalogowy: (1535857.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
j.m.: 1 * 1 SZT
Numer katalogowy: (1547153.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
j.m.: 1 * 200 mg
Numer katalogowy: (1707931.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
j.m.: 1 * 1 g
Numer katalogowy: (1309911.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
j.m.: 1 * 1 g
Numer katalogowy: (1270231.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
j.m.: 1 * 1 g
Numer katalogowy: (1535846.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
j.m.: 1 * 1 SZT
Numer katalogowy: (1535813.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
j.m.: 1 * 1 SZT
Numer katalogowy: (1609840.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
j.m.: 1 * 1 g
Numer katalogowy: (1672100.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
j.m.: 1 * 1 g
Numer katalogowy: (1043309.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the F...
j.m.: 1 * 1 g
Cena wymaga potwierdzenia u dostawcy
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Ten produkt jest ograniczony w dostępie i można go zamówić tylko przy użyciu zaaprobowanego konta. Jeśli potrzebujesz pomocy, napisz do Działu Prawnego VWR na tomasz.chrobak@pl.vwr.com
Do zakupu tego produktu może być potrzebna dodatkowa dokumentacja. Przedstawiciel VWR skontaktuje się z Tobą w razie potrzeby.
Ten produkt jest zablokowany na stronie internetowej. W celu zamówienia, skontaktuj się z Działem Obsługi Klienta.
Produkt oryginalny został wycofany. Dostępny jest wskazny zamiennik .
Wybrany produkt został wycofany - sprzedaż do wyczerpania zapasów. Dostępne zamienniki można znaleźć poprzez wpisanie powyższego numeru katalogowego VWR w okno wyszukiwarki. Jeśli potrzebujesz dalszej pomocy, prosimy o kontakt z Działem Doradztwa Produktowego pod numerem telefonu 58 3238 220.
1 - 16 of 125
no targeter for Bottom