Temperature loggers, CurveX-3 basic oven logger
- Catalog No:
- SIMECX3010
- SIMECX3005
- Display:
- Three multi-colours LED's
- Three multi-colours LED's
- Interface:
- USB
- USB
- Battery life:
- 1200 hour continuous use
- 1200 hour continuous use
- Recording intervals:
- 1 s - 60 min
- 1 s - 60 min
- Weight:
- 190 g
- 190 g
- W×D×H:
- 100×85×16 mm
- 100×85×16 mm
- Data memory - No. of readings:
- 10 batches with 16000 or 1 batch with 160000 readings
- 10 batches with 16000 or 1 batch with 160000 readings
Product Information
- Katalog #Dostawca #RangeAccuracyPkAvailabilityPrice
- Oven loggers, CurveX 3 BasicOven loggers, CurveX 3 BasicSIMECX3005CX3005T: 0...+500 °C/−58...−932 °FT: ±1 °C11 * 1 SZT7 150 złSupplier Article Number: CX3005Local Article Number: SIMECX3005
Accessories
Specifications
Environmentally PreferableDisplayThree multi-colours LED'sInterfaceUSBBattery life1200 hour continuous useRecording intervals1 s - 60 minWeight190 gW×D×H100×85×16 mmData memory - No. of readings10 batches with 16000 or 1 batch with 160000 readings - Oven logger kit, CurveX 3 Basic, incl. insulation box and insulation box logger bracketOven logger kit, CurveX 3 Basic, incl. insulation box and insulation box logger bracketSIMECX3010CX3010T: 0...+500 °CT: ±1 °C11 * 1 SZT14 920 złSupplier Article Number: CX3010Local Article Number: SIMECX3010
Accessories
Specifications
Environmentally PreferableDisplayThree multi-colours LED'sInterfaceUSBBattery life1200 hour continuous useRecording intervals1 s - 60 minWeight190 gW×D×H100×85×16 mmData memory - No. of readings10 batches with 16000 or 1 batch with 160000 readings
Probes
- SeqDescriptionRangePk
- 28Air/Surface probe, wire type, inconel tube 3 m cable0…1000 °C1: CX2094: SIMECX2094