Finnpipette™ F1 Multi-Channel Pipettes, Mechanical, Variable Volume
Supplier: Thermo Fisher Scientific
Certificates
About this item
Limited Time Sale
Select items below are currently on sale
Specifications
- Catalog No:
- 613-5576
- 613-5577
- 613-5574
- 613-5575
- 613-5569
- 613-5570
- 613-5568
- 613-5572
- 613-5573
- 613-5571
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
Specifications
8-Channel
Product Information
- Catalog #Supplier #VolumeIncrementAccuracyImprecisionColour codeAvailabilityPrice
- Finnpipette F1Finnpipette F1613-55684661000N1 - 10 µl0,02 µl±12,00 - ±2,40 %8,00 - 1,60 %PinkLog in for availability1 * 1 SZT1 * 1 SZT - 2 676 zł2 676 zł
Sale Valid until 30/06/2025
Sale Valid until 30/06/20251 * 1 SZT2 676 złSpecifications:Low Manufacturing Impact Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Finnpipette F1Finnpipette F1613-55694661010N5 - 50 µl0,1 µl±5,00 - ±1,50 %2,00 - 0,70 %YellowLog in for availability1 * 1 SZT1 * 1 SZT - 2 676 zł2 676 zł
Sale Valid until 30/06/2025
Sale Valid until 30/06/20251 * 1 SZT2 676 złSpecifications:Low Manufacturing Impact Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Finnpipette F1Finnpipette F1613-55704661020N10 - 100 µl0,2 µl±5,00 - ±1,30 %2,00 - 0,50 %YellowLog in for availability1 * 1 SZT1 * 1 SZT - 2 676 zł2 676 zł
Sale Valid until 30/06/2025
Sale Valid until 30/06/20251 * 1 SZT2 676 złSpecifications:Low Manufacturing Impact Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Finnpipette F1Finnpipette F1613-55714661030N30 - 300 µl1 µl±5,00 - ±1,00 %2,00 - 0,30 %OrangeLog in for availability1 * 1 SZT1 * 1 SZT - 2 676 zł2 676 zł
Sale Valid until 30/06/2025
Sale Valid until 30/06/20251 * 1 SZT2 676 złSpecifications:Low Manufacturing Impact Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
12-Channel
Product Information
- Catalog #Supplier #VolumeIncrementAccuracyImprecisionColour codeAvailabilityPrice
- Finnpipette F1Finnpipette F1613-55744661060N10 - 100 µl0,2 µl±5,00 - ±1,30 %2,00 - 0,50 %YellowLog in for availability1 * 1 SZT1 * 1 SZT - 3 032 zł3 032 zł
Sale Valid until 30/06/2025
Sale Valid until 30/06/20251 * 1 SZT3 032 złSpecifications:Low Manufacturing Impact Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Finnpipette F1Finnpipette F1613-55754661070N30 - 300 µl1 µl±5,00 - ±1,00 %2,00 - 0,30 %OrangeLog in for availability1 * 1 SZT1 * 1 SZT - 3 032 zł3 032 zł
Sale Valid until 30/06/2025
Sale Valid until 30/06/20251 * 1 SZT3 032 złSpecifications:Low Manufacturing Impact Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Finnpipette F1Finnpipette F1613-55724661040N1 - 10 µl0,02 µl±12,00 - ±2,40 %8,00 - 1,60 %PinkLog in for availability1 * 1 SZT1 * 1 SZT - 3 032 zł3 032 zł
Sale Valid until 30/06/2025
Sale Valid until 30/06/20251 * 1 SZT3 032 złSpecifications:Low Manufacturing Impact Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Finnpipette F1Finnpipette F1613-55734661050N5 - 50 µl0,1 µl±5,00 - ±1,50 %2,00 - 0,70 %YellowLog in for availability1 * 1 SZT1 * 1 SZT - 3 032 zł3 032 zł
Sale Valid until 30/06/2025
Sale Valid until 30/06/20251 * 1 SZT3 032 złSpecifications:Low Manufacturing Impact Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
16-Channel
Product Information
- Catalog #Supplier #VolumeIncrementAccuracyImprecisionColour codeAvailabilityPrice
- Finnpipette F1Finnpipette F1613-55764661080N1 - 10 µl0,02 µl±12,00 - ±2,40 %8,00 - 1,60 %PinkLog in for availability1 * 1 SZT1 * 1 SZT - 3 245 zł3 245 zł
Sale Valid until 30/06/2025
Sale Valid until 30/06/20251 * 1 SZT3 245 złSpecifications:Low Manufacturing Impact Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - Finnpipette F1Finnpipette F1613-55774661090N5 - 50 µl0,1 µl±5,00 - ±1,50 %2,00 - 0,70 %TurquoiseLog in for availability1 * 1 SZT1 * 1 SZT - 3 245 zł3 245 zł
Sale Valid until 30/06/2025
Sale Valid until 30/06/20251 * 1 SZT3 245 złSpecifications:Low Manufacturing Impact Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact