Drukuj
Drabinka DNA, 100 bp plus
Dostawca: PanReac AppliChem
A5216.0005EA 0 PLN
APLIA5216.0005 APLIA5216.0025
Drabinka DNA, 100 bp plus
This 100 bp DNA Ladder is ideal for determining the size of double-stranded DNA from 100 to 1,500 base pairs. The ladder consists of ten fragments ranging in size from 100 - 1000 bp in 100 bp increments plus one additional 1500 bp fragment. The 500 bp and 1000 bp fragment are present at increased intensity to allow easier identification. All fragments are blunt-ended. The fragment sizes (in base pairs) are 1500, 1000 (×2), 900, 800, 700, 600, 500 (×2), 400, 300, 200, 100.
The DNA ladder is supplied at 0,2 mg/ml in 10 mM Tris · HCl (pH 8,0), 1,0 mM EDTA. The recommended amount of size marker to load on the gel is 0.4 to 0,6 μg (2 to 3 μl) per lane.