Drukuj
Drabinka DNA, 1 kb
Dostawca: Applichem Lifescience
A5207.0025EA 0 PLN
A5207.0025 A5207.0005
Drabinka DNA, 1 kb
Elektroforezy, Odczynniki Markery masy cząsteczkowej
This 1kb DNA ladder is a convenient marker for determining the size of double-stranded DNA from 250 to 10,000 base pairs. The ladder consists of 13 fragments ranging in size from 250 to 10,000 base pairs. The 1000 bp and 3000 bp fragments have an increased intensity relative to the other bands on ethidium bromide-stained agarose gels and serve as reference indicators. All fragments are blunt-ended. The exact fragment sizes (in kb) are 10,0, 8,0, 6,0, 5,0, 4,0, 3,0 (×2), 2,5, 2,0, 1,5, 1,0 (×2), 0,75, 0,5, 0,25.
The marker is supplied in the storage buffer containing 10 mM Tris · HCl (pH 7.8), 10 mM NaCl, 1 mM EDTA at a concentration of 0,2 mg/ml).