Order Entry
VWR® ARISTAR®, Cerium Standard Solutions for ICP-MS
VWR® ARISTAR®, Cerium Standard Solutions for ICP-MS
undefined
VWR® ARISTAR®, Cerium Standard Solutions for ICP-MS
  •  
    Element
    Symbol
    Concentration
    Matrix
    Pack type
    Pk
  • 85557.180
    Cerium
    Ce
    10 mg/L
    2% HNO₃
    Plastic bottle
    100 mL
    1 * 100 mL
    1 * 100 mL - 758 zł
    758 zł
    1 * 100 mL
       758 zł
     : VWRC85557.180
  • 457224C
    Cerium
    Ce
    1000 mg/L
    2-5% HNO₃
    Plastic bottle
    100 mL
    1 * 100 mL
    1 * 100 mL - 761 zł
    761 zł
    1 * 100 mL
       761 zł
     : VWRC457224C