About this item
Recognizes a 100 kDa glycoprotein, identified as CD10, also known as Common Acute Lymphocytic Leukemia Antigen (CALLA). It is a cell surface enzyme with neutral metalloendopeptidase activity, which inactivates a variety of biologically active peptides. CD10 is expressed on the cells of lymphoblastic, Burkitt's, and follicular germinal center lymphomas, and on cells from patients with chronic myelocytic leukemia (CML). It is also expressed on the surface of normal early lymphoid progenitor cells, immature B cells within adult bone marrow and germinal center B cells within lymphoid tissue. CD10 is also present on breast myoepithelial cells, bile canaliculi, fibroblasts, with especially high expression on the brush border of kidney and gut epithelial cells.
Type: Primary
Antigen: MME
Clonality: Monoclonal
Clone: CB-CALLA
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Specifications
- Catalog No:
- BNC680146-500
- BNC680146-100
- Antigen symbol:
- MME
- MME
- Antigen name:
- CD10
- CD10
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 568
- CF® 568
- Clone:
- Clone CB-CALLA
- Clone CB-CALLA
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 307734
- 307734
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Product Information
- Catalog #Supplier #SizePkAvailabilityPrice
- Specifications:
More
Specifications
Antigen symbolMMEAntigen nameCD10Environmentally PreferableAntibody typePrimaryClonalityMonoclonalConjugationCF® 568CloneClone CB-CALLAReactivityHumanHostMouseGene ID307734IsotypeIgG1 kappaImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide - Specifications:
More
Specifications
Antigen symbolMMEAntigen nameCD10Environmentally PreferableAntibody typePrimaryClonalityMonoclonalConjugationCF® 568CloneClone CB-CALLAReactivityHumanHostMouseGene ID307734IsotypeIgG1 kappaImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide