Order Entry
Anti-GFAP Mouse Monoclonal Antibody (CF488A) [clone: GA-5]
Anti-GFAP Mouse Monoclonal Antibody (CF488A) [clone: GA-5]
 :  Biotium
Anti-GFAP Mouse Monoclonal Antibody (CF488A) [clone: GA-5]
 :  Biotium
  • Catalog No:
  • BNC880167-500
  • BNC880167-100
  • Antigen symbol:
  • GFAP
  • GFAP
  • Antigen name:
  • Glial fibrillary acidic protein
  • Glial fibrillary acidic protein
  • Antigen synonyms:
  • Astrocyte or Intermediate Filament Protein|Glial Fibrillary Acidic Protein (GFAP)
  • Astrocyte or Intermediate Filament Protein|Glial Fibrillary Acidic Protein (GFAP)
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone GA-5
  • Clone GA-5
  • Reactivity:
  • Pig, Rabbit, Chicken, Human, Bovine, Rat, Mouse
  • Pig, Rabbit, Chicken, Human, Bovine, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 514227
  • 514227
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide
  •  
    Size
    Pk
  • BNC880167-500
    BNC880167-500
    500 μL
    500 µl
    1 * 500 µl
    1 * 500 µl - 2 860 zł
    2 860 zł
    1 * 500 µl
       2 860 zł
    Specifications
    Antigen symbol
    GFAP
    Antigen name
    Glial fibrillary acidic protein
    Environmentally Preferable
    Antigen synonyms
    Astrocyte or Intermediate Filament Protein|Glial Fibrillary Acidic Protein (GFAP)
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 488A
    Clone
    Clone GA-5
    Reactivity
    Pig, Rabbit, Chicken, Human, Bovine, Rat, Mouse
    Host
    Mouse
    Gene ID
    514227
    Isotype
    IgG1
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide
  • BNC880167-100
    BNC880167-100
    100 μL
    100 µl
    1 * 100 µl
    1 * 100 µl - 1 040 zł
    1 040 zł
    1 * 100 µl
       1 040 zł
    Specifications
    Antigen symbol
    GFAP
    Antigen name
    Glial fibrillary acidic protein
    Environmentally Preferable
    Antigen synonyms
    Astrocyte or Intermediate Filament Protein|Glial Fibrillary Acidic Protein (GFAP)
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 488A
    Clone
    Clone GA-5
    Reactivity
    Pig, Rabbit, Chicken, Human, Bovine, Rat, Mouse
    Host
    Mouse
    Gene ID
    514227
    Isotype
    IgG1
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide