Order Entry
Anti-MSN Mouse Monoclonal Antibody (CF568) [clone: MSN492]
Anti-MSN Mouse Monoclonal Antibody (CF568) [clone: MSN492]
 :  Biotium
Anti-MSN Mouse Monoclonal Antibody (CF568) [clone: MSN492]
 :  Biotium
  • Catalog No:
  • BNC680492-500
  • BNC680492-100
  • Antigen symbol:
  • MSN
  • MSN
  • Antigen name:
  • Moesin
  • Moesin
  • Antigen synonyms:
  • MSN/ALK fusion|Membrane-organizing extension spike protein|Moesin/anaplastic lymphoma kinase fusion protein
  • MSN/ALK fusion|Membrane-organizing extension spike protein|Moesin/anaplastic lymphoma kinase fusion protein
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • Clone MSN492
  • Clone MSN492
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 87752
  • 87752
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide
  •  
    Size
    Pk
  • BNC680492-500
    BNC680492-500
    500 μL
    500 µl
    1 * 500 µl
    1 * 500 µl - 2 860 zł
    2 860 zł
    1 * 500 µl
       2 860 zł
     : BNC680492-500
     : BTIUBNC680492-500
    Specifications
    Antigen symbol
    MSN
    Antigen name
    Moesin
    Environmentally Preferable
    Antigen synonyms
    MSN/ALK fusion|Membrane-organizing extension spike protein|Moesin/anaplastic lymphoma kinase fusion protein
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 568
    Clone
    Clone MSN492
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    87752
    Isotype
    IgG1 kappa
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide
  • BNC680492-100
    BNC680492-100
    100 μL
    100 µl
    1 * 100 µl
    1 * 100 µl - 1 040 zł
    1 040 zł
    1 * 100 µl
       1 040 zł
     : BNC680492-100
     : BTIUBNC680492-100
    Specifications
    Antigen symbol
    MSN
    Antigen name
    Moesin
    Environmentally Preferable
    Antigen synonyms
    MSN/ALK fusion|Membrane-organizing extension spike protein|Moesin/anaplastic lymphoma kinase fusion protein
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 568
    Clone
    Clone MSN492
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    87752
    Isotype
    IgG1 kappa
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide