Order Entry
Anti-KRT10 Mouse Monoclonal Antibody (CF594) [clone: LH2]
Anti-KRT10 Mouse Monoclonal Antibody (CF594) [clone: LH2]
Supplier:  Biotium
Certificates
Anti-KRT10 Mouse Monoclonal Antibody (CF594) [clone: LH2]
Supplier:  Biotium
Specifications
  • Catalog No:
  • BNC940674-500
  • BNC940674-100
  • Antigen symbol:
  • KRT10
  • KRT10
  • Antigen name:
  • Cytokeratin 10
  • Cytokeratin 10
  • Antigen synonyms:
  • Keratin Type I Cytoskeletal 10|EHK|BIE|BCIE|KRT10
  • Keratin Type I Cytoskeletal 10|EHK|BIE|BCIE|KRT10
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Clone:
  • Clone LH2
  • Clone LH2
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 99936
  • 99936
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide
Specifications
Product Information
  • Catalog #
    Supplier #
    Size
    Pk
    Availability
    Price
  • BNC940674-500
    BNC940674-500
    500 μL
    500 µl
    Log in for availability
    Log in for availability
    1 * 500 µl
    1 * 500 µl - 2 860 zł
    2 860 zł
    1 * 500 µl
       2 860 zł
    Specifications:
    More
    Specifications
    Antigen symbol
    KRT10
    Antigen name
    Cytokeratin 10
    Environmentally Preferable
    Antigen synonyms
    Keratin Type I Cytoskeletal 10|EHK|BIE|BCIE|KRT10
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 594
    Clone
    Clone LH2
    Reactivity
    Human, Mouse
    Host
    Mouse
    Gene ID
    99936
    Isotype
    IgG1 kappa
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide
  • BNC940674-100
    BNC940674-100
    100 μL
    100 µl
    Log in for availability
    Log in for availability
    1 * 100 µl
    1 * 100 µl - 1 040 zł
    1 040 zł
    1 * 100 µl
       1 040 zł
    Specifications:
    More
    Specifications
    Antigen symbol
    KRT10
    Antigen name
    Cytokeratin 10
    Environmentally Preferable
    Antigen synonyms
    Keratin Type I Cytoskeletal 10|EHK|BIE|BCIE|KRT10
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 594
    Clone
    Clone LH2
    Reactivity
    Human, Mouse
    Host
    Mouse
    Gene ID
    99936
    Isotype
    IgG1 kappa
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide