Anti-IGHM Mouse Monoclonal Antibody (Purified) [clone: DA4-4]
Supplier: Biotium
Certificates
About this item
Recognizes a protein of 75 kDa, identified as mu heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), gamma (IgG), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This MAb is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.
Type: Primary
Antigen: IGHM
Clonality: Monoclonal
Clone: DA4-4
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- BNUB0759-100
- BNUB0759-500
- Antigen symbol:
- IGHM
- IGHM
- Antigen name:
- IgM Immunoglobulin
- IgM Immunoglobulin
- Antigen synonyms:
- Ig Mu Chain C Region|Constant Region of Heavy Chain of IgM|AGM1|IGHM
- Ig Mu Chain C Region|Constant Region of Heavy Chain of IgM|AGM1|IGHM
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- Clone DA4-4
- Clone DA4-4
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 510635
- 510635
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,2 mg/ml
- 0,2 mg/ml
- Storage buffer:
- PBS/0,05% BSA/0,05% azide
- PBS/0,05% BSA/0,05% azide
Specifications
Product Information
- Catalog #Supplier #SizePkAvailabilityPrice
- Supplier Article Number: BNUB0759-100Local Article Number: BTIUBNUB0759-100Specifications:
More
Specifications
Antigen symbolIGHMAntigen nameIgM ImmunoglobulinEnvironmentally PreferableAntigen synonymsIg Mu Chain C Region|Constant Region of Heavy Chain of IgM|AGM1|IGHMAntibody typePrimaryClonalityMonoclonalConjugationUnconjugatedCloneClone DA4-4ReactivityHumanHostMouseGene ID510635IsotypeIgG1 kappaImmunoChemistryYesImmunoFluorescenceYesConcentration0,2 mg/mlStorage bufferPBS/0,05% BSA/0,05% azide - Supplier Article Number: BNUB0759-500Local Article Number: BTIUBNUB0759-500Specifications:
More
Specifications
Antigen symbolIGHMAntigen nameIgM ImmunoglobulinEnvironmentally PreferableAntigen synonymsIg Mu Chain C Region|Constant Region of Heavy Chain of IgM|AGM1|IGHMAntibody typePrimaryClonalityMonoclonalConjugationUnconjugatedCloneClone DA4-4ReactivityHumanHostMouseGene ID510635IsotypeIgG1 kappaImmunoChemistryYesImmunoFluorescenceYesConcentration0,2 mg/mlStorage bufferPBS/0,05% BSA/0,05% azide