About this item
This MAb recognizes an epitope within the 74-182 C-terminal sequence (11kD peptide fragment) of human serum Cellular Retinol Binding Protein 1 (CRBP 1), a single-chain glycoprotein belonging to the superfamily of hydrophobic molecule transporter proteins, which is responsible for transport of retinol (vitamin A1) from the liver to peripheral target tissues, like the eye, where it mediates the cellular uptake. CRBP 1 is synthesized by hepatic parenchymal cells where it becomes bound to its ligand retinol and is then released into the circulation, where it binds further to the protein transthyretin, to form a transporting complex, which is big enough not to be lost by filtration through the kidney glomeruli. It is detected in nearly all tissues with higher expression in adult ovary, pancreas, pituitary gland, adrenal gland, and fetal liver.
Type: Primary
Antigen: RBP1
Clonality: Monoclonal
Clone: G4E4
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Chimpanzee, Goat, Human, Monkey, Mouse, Rabbit, Rat
Specifications
- Catalog No:
- BNCB0855-100
- BNCB0855-500
- Antigen symbol:
- RBP1
- RBP1
- Antigen name:
- Retinol binding protein 1
- Retinol binding protein 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Clone:
- Clone G4E4
- Clone G4E4
- Reactivity:
- Rabbit, Monkey, Goat, Chimpanzee, Human, Rat, Mouse
- Rabbit, Monkey, Goat, Chimpanzee, Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 529571
- 529571
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Product Information
- Catalog #Supplier #SizePkAvailabilityPrice
- Supplier Article Number: BNCB0855-100Local Article Number: BTIUBNCB0855-100Specifications:
More
Specifications
Antigen symbolRBP1Antigen nameRetinol binding protein 1Environmentally PreferableAntibody typePrimaryClonalityMonoclonalConjugationBiotinCloneClone G4E4ReactivityRabbit, Monkey, Goat, Chimpanzee, Human, Rat, MouseHostMouseGene ID529571IsotypeIgG1 kappaImmunoChemistryYesImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide - Supplier Article Number: BNCB0855-500Local Article Number: BTIUBNCB0855-500Specifications:
More
Specifications
Antigen symbolRBP1Antigen nameRetinol binding protein 1Environmentally PreferableAntibody typePrimaryClonalityMonoclonalConjugationBiotinCloneClone G4E4ReactivityRabbit, Monkey, Goat, Chimpanzee, Human, Rat, MouseHostMouseGene ID529571IsotypeIgG1 kappaImmunoChemistryYesImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide