Poland
Order Entry
Anti-CYCS Mouse Monoclonal Antibody (CF568) [clone: CYCS/1010]
Anti-CYCS Mouse Monoclonal Antibody (CF568) [clone: CYCS/1010]
Supplier:  Biotium
Certificates
Anti-CYCS Mouse Monoclonal Antibody (CF568) [clone: CYCS/1010]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC681010-500
  • BNC681010-100
  • Antigen symbol:
  • CYCS
  • CYCS
  • Antigen name:
  • Cytochrome C
  • Cytochrome C
  • Antigen synonyms:
  • HCS|THC4|CYCS|CYC
  • HCS|THC4|CYCS|CYC
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • Clone CYCS/1010
  • Clone CYCS/1010
  • Reactivity:
  • Human, Rat
  • Human, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 437060
  • 437060
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Product Information

  • Catalog #
    Supplier #
    Size
    Pk
    Availability
    Price
  • BNC681010-500
    BNC681010-500
    500 μL
    500 µl
    Log in for availability
    Log in for availability
    1 * 500 µl
    1 * 500 µl - 2 860 zł
    2 860 zł
    1 * 500 µl
       2 860 zł

    Supplier Article Number: BNC681010-500
    Local Article Number: BTIUBNC681010-500
    Specifications:
    More
    Specifications
    Antigen symbol
    CYCS
    Antigen name
    Cytochrome C
    Environmentally Preferable
    Antigen synonyms
    HCS|THC4|CYCS|CYC
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 568
    Clone
    Clone CYCS/1010
    Reactivity
    Human, Rat
    Host
    Mouse
    Gene ID
    437060
    Isotype
    IgG2b kappa
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide
  • BNC681010-100
    BNC681010-100
    100 μL
    100 µl
    Log in for availability
    Log in for availability
    1 * 100 µl
    1 * 100 µl - 1 040 zł
    1 040 zł
    1 * 100 µl
       1 040 zł

    Supplier Article Number: BNC681010-100
    Local Article Number: BTIUBNC681010-100
    Specifications:
    More
    Specifications
    Antigen symbol
    CYCS
    Antigen name
    Cytochrome C
    Environmentally Preferable
    Antigen synonyms
    HCS|THC4|CYCS|CYC
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 568
    Clone
    Clone CYCS/1010
    Reactivity
    Human, Rat
    Host
    Mouse
    Gene ID
    437060
    Isotype
    IgG2b kappa
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide