About this item
This MAb reacts with the beta-chain of HLA-DRB1 antigen, a member of MHC class II molecules. It does not cross react with HLA-DP and HLA-DQ. HLA-DR is a heterodimeric cell surface glycoprotein comprised of a 36 kDa alpha (heavy) chain and a 28 kDa beta (light) chain. It is expressed on B-cells, activated T-cells, monocytes/macrophages, dendritic cells and other non-professional APCs. In conjunction with the CD3/TCR complex and CD4 molecules, HLA-DR is critical for efficient peptide presentation to CD4 T cells. It is an excellent histiocytic marker in paraffin sections producing intense cytoplasmic staining. True histiocytic neoplasms are similarly positive. HLA-DR antigens also occur on a variety of epithelial cells and their corresponding neoplastic counterparts. Loss of HLA-DR expression is related to tumor microenvironment and predicts adverse outcome in diffuse large B-cell lymphoma.
Type: Primary
Antigen: HLA-DRB1
Clonality: Monoclonal
Clone: LN-3 HLA-DRB/1067
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Monkey
Specifications
- Catalog No:
- BNC041208-100
- BNC041208-500
- Antigen symbol:
- HLA-DRB1
- HLA-DRB1
- Antigen name:
- HLA-DRB
- HLA-DRB
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 405S
- CF® 405S
- Clone:
- Clone LN-3 HLA-DRB/1067
- Clone LN-3 HLA-DRB/1067
- Reactivity:
- Monkey, Human
- Monkey, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 534322
- 534322
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Product Information
- Catalog #Supplier #SizePkAvailabilityPrice
- Specifications:
More
Specifications
Antigen symbolHLA-DRB1Antigen nameHLA-DRBEnvironmentally PreferableAntibody typePrimaryClonalityMonoclonalConjugationCF® 405SCloneClone LN-3 HLA-DRB/1067ReactivityMonkey, HumanHostMouseGene ID534322IsotypeIgGWestern blotYesImmunoChemistryYesImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide - Specifications:
More
Specifications
Antigen symbolHLA-DRB1Antigen nameHLA-DRBEnvironmentally PreferableAntibody typePrimaryClonalityMonoclonalConjugationCF® 405SCloneClone LN-3 HLA-DRB/1067ReactivityMonkey, HumanHostMouseGene ID534322IsotypeIgGWestern blotYesImmunoChemistryYesImmunoFluorescenceYesConcentration0,1 mg/mlStorage bufferPBS/0,1% BSA/0,05% azide