Poland
Złóż zamówienie
Agaroza, SeaKem® Gold
Agaroza, SeaKem® Gold
Supplier:  LONZA
Certificates
undefined
Agaroza, SeaKem® Gold
Supplier:  LONZA
Product Information
  • Catalog #
    Supplier #
    Opis
    Ilość
    Availability
    Price
  • 733-1552
    50152
    Agarose, SeaKem® Gold
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 2 440 zł
    2 440 zł
    1 * 25 g
       2 440 zł
    Supplier Article Number: 50152
    Local Article Number: LONZ50152
    Specifications:
    Specifications
    Environmentally Preferable
  • 733-1551
    50150
    Agarose, SeaKem® Gold
    125 g
    Log in for availability
    Log in for availability
    1 * 125 g
    1 * 125 g - 8 160 zł
    8 160 zł
    1 * 125 g
       8 160 zł
    Supplier Article Number: 50150
    Local Article Number: LONZ50150
    Specifications:
    Specifications
    Environmentally Preferable