Qualitative filter papers, wet strengthened grades, grade 91, Whatman™
Supplier: Whatman products (Cytiva)
Certificates
About this item
A general purpose creped cellulose filter for less critical routine analysis. Widely used to assay sucrose in cane sugar and within pharmaceutical laboratories for routine filtration.
These extremely strong filter papers have a high wet strength due to the addition of a small quantity of chemically stable resin. Normal qualitative applications will not introduce any significant impurities into the filtrate. The resins do, however, contain nitrogen so these grades should not be used in Kjeldahl estimations, etc.
Specifications
- Catalog No:
- 512-0386
- 512-0388
- 512-0387
- 512-0613
- 512-0389
- 512-0612
- 516-0604
- 512-0648
- 512-0614
- Grade:
- 91
- 91
- 91
- 91
- 91
- 91
- 91
- 91
- 91
- Weight:
- 65 g/m²
- 65 g/m²
- 65 g/m²
- 65 g/m²
- 65 g/m²
- 65 g/m²
- 65 g/m²
- 65 g/m²
- 65 g/m²
- Thickness:
- 205 µm
- 205 µm
- 205 µm
- 205 µm
- 205 µm
- 205 µm
- 205 µm
- 205 µm
- 205 µm
- Particle retention:
- 10 µm
- 10 µm
- 10 µm
- 10 µm
- 10 µm
- 10 µm
- 10 µm
- 10 µm
- 10 µm
- Filtration speed:
- 70 s (Herzberg)
- 70 s (Herzberg)
- 70 s (Herzberg)
- 70 s (Herzberg)
- 70 s (Herzberg)
- 70 s (Herzberg)
- 70 s (Herzberg)
- 70 s (Herzberg)
- 70 s (Herzberg)
- Airflow rate:
- 6 s/100 ml/6,5 cm²
- 6 s/100 ml/6,5 cm²
- 6 s/100 ml/6,5 cm²
- 6 s/100 ml/6,5 cm²
- 6 s/100 ml/6,5 cm²
- 6 s/100 ml/6,5 cm²
- 6 s/100 ml/6,5 cm²
- 6 s/100 ml/6,5 cm²
- 6 s/100 ml/6,5 cm²
Specifications
Product Information
- Catalog #Supplier #GradeSizePkAvailabilityPrice
- Specifications:
More
Specifications
Grade91Environmentally PreferableWeight65 g/m²Thickness205 µmParticle retention10 µmFiltration speed70 s (Herzberg)Airflow rate6 s/100 ml/6,5 cm² - Specifications:
More
Specifications
Grade91Environmentally PreferableWeight65 g/m²Thickness205 µmParticle retention10 µmFiltration speed70 s (Herzberg)Airflow rate6 s/100 ml/6,5 cm² - Specifications:
More
Specifications
Grade91Environmentally PreferableWeight65 g/m²Thickness205 µmParticle retention10 µmFiltration speed70 s (Herzberg)Airflow rate6 s/100 ml/6,5 cm² - Specifications:
More
Specifications
Grade91Environmentally PreferableWeight65 g/m²Thickness205 µmParticle retention10 µmFiltration speed70 s (Herzberg)Airflow rate6 s/100 ml/6,5 cm² - Specifications:
More
Specifications
Grade91Environmentally PreferableWeight65 g/m²Thickness205 µmParticle retention10 µmFiltration speed70 s (Herzberg)Airflow rate6 s/100 ml/6,5 cm² - Specifications:
More
Specifications
Grade91Environmentally PreferableWeight65 g/m²Thickness205 µmParticle retention10 µmFiltration speed70 s (Herzberg)Airflow rate6 s/100 ml/6,5 cm² - Specifications:
More
Specifications
Grade91Environmentally PreferableWeight65 g/m²Thickness205 µmParticle retention10 µmFiltration speed70 s (Herzberg)Airflow rate6 s/100 ml/6,5 cm² - Specifications:
More
Specifications
Grade91Environmentally PreferableWeight65 g/m²Thickness205 µmParticle retention10 µmFiltration speed70 s (Herzberg)Airflow rate6 s/100 ml/6,5 cm² - Specifications:
More
Specifications
Grade91Environmentally PreferableWeight65 g/m²Thickness205 µmParticle retention10 µmFiltration speed70 s (Herzberg)Airflow rate6 s/100 ml/6,5 cm²