Order Entry
Pig Pepsin (from Stomach mucosa), MP Biomedicals
Pig Pepsin (from Stomach mucosa), MP Biomedicals
Pig Pepsin (from Stomach mucosa), MP Biomedicals
  • CAS No.:
    9001-75-6
  • Source:
    Stomach mucosa
  • Species:
    Pig
  • Storage conditions:
    −20 °C
  • Form:
    Yellow lyophilized powder
  • Enzyme name:
    Pepsin
  • Enzyme activity:
    >2000 u/mg protein
  • Molecular weight:
    35 kDa
  • pH:
    1,0 - 3,5
  •  
    Pk
  • ICNA02195367.2
    02195367.2
    250 mg
    1 * 250 mg
    1 * 250 mg - 311 zł
    311 zł
    1 * 250 mg
       311 zł
     : 02195367.2
     : ICNA02195367.2
  • ICNA0219536701
    0219536701
    1 g
    1 * 1 g
    1 * 1 g - 565 zł
    565 zł
    1 * 1 g
       565 zł
     : 0219536701
     : ICNA0219536701
  • ICNA0219536705
    0219536705
    5 g
    1 * 5 g
    1 * 5 g - 2 440 zł
    2 440 zł
    1 * 5 g
       2 440 zł
     : 0219536705
     : ICNA0219536705
  • ICNA0219536710
    0219536710
    10 g
    1 * 10 g
    1 * 10 g - 3 980 zł
    3 980 zł
    1 * 10 g
       3 980 zł
     : 0219536710
     : ICNA0219536710