Drukuj
Cyrkulatory chłodzące
Dostawca: Huber
461-0145EA 0 PLN
461-0145 461-0564 461-0142 461-0143
Cyrkulatory chłodzące
Termostaty cyrkulacyjne Systemy cyrkulacyjne, otwarte Łaźnie ze zdejmowanym cyrkulatorem
These circulators, with insulated refrigeration baths, are designed for direct temperature control from −30 up to +200 °C.
  • Pilot ONE® is interchangeable and can be used as a remote control (with an optional data cable)
  • Controller can be upgraded with enhanced functionality at any time (Exclusive and Professional E-grade® packages), basic version features six languages (English, German, French, Spanish, Italian and Russian)
  • The display resolution is 0,1 °C; display can be customised to the users' preferences and important information e.g. set point, actual internal or process temperatures can be displayed in a larger format
  • Powerful variable speed pressure/suction pump with Variable Pressure Control (VPC)
  • User can set visual and acoustic alarms, limiting the temperature range, and program the unit with its clock and calendar functions for auto start for unsupervised start-up
The Pilot ONE® controller offers a brilliant 5,7" touch screen with graphic functions and a comfortable navigation menu in 11 languages and USB and Ethernet connections.
Maximum pump capacities:
Pilot ONE® (CC-E): 27 L/min, 700 mbar, 25 L/min, 400 mbar (suction)
Certyfikaty: Comply with DIN 12876 Safety class III, FL
Uwaga: *Cooling capacity at 0; −10 and −20 °C