Yeast aldehyde dehydrogenase, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
- From Yeast Potassium Activated (Aldehyde: NAD + (P+) ov. reductase)
- E.C.1.2.1.5
- Freeze dried stabilized powder
- Activity: 2 unit/mg solid (approx. 5 u/mg protein)
- ADH Less than 0,1% Unit
Definition: One unit causes the oxidation of one micromole of acetaldehyde to acetic acid per minute at pH 8,0 and 25 °C in the presence of potassium ions and thiols.
Specifications
- CAS No.:9028-86-8
- Species:Yeast
- Storage conditions:−20 °C
- Enzyme name:Aldehyde Dehydrogenase
- Molecular weight:~200000 (Lit.)
Specifications
Product Information
- Catalog #Supplier #PkAvailabilityPrice
- Supplier Article Number: 0219067925Local Article Number: ICNA0219067925
- Supplier Article Number: 0219067980Local Article Number: ICNA0219067980
- Supplier Article Number: 0219067983Local Article Number: ICNA0219067983