Drukuj

Anti-ITGA3 Rabbit Polyclonal Antibody (Cy5®)

Dostawca: Bioss
BOSSBS-2093R-CY5EA 4310 PLN
BOSSBS-2093R-CY5
Anti-ITGA3 Rabbit Polyclonal Antibody (Cy5®)
Przeciwciała
Integrin alpha-3/beta-1 is a receptor for fibronectin, laminin, collagen, epiligrin, thrombospondin and CSPG4. Integrin alpha-3/beta-1 provides a docking site for FAP (seprase) at invadopodia plasma membranes in a collagen-dependent manner and hence may participate in the adhesion, formation of invadopodia and matrix degradation processes, promoting cell invasion. Alpha-3/beta-1 may mediate with LGALS3 the stimulation by CSPG4 of endothelial cells migration.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ITGA3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR