Drukuj

Anti-TNFSF18 Rabbit Polyclonal Antibody (Cy5.5®)

Dostawca: Bioss
BOSSBS-2456R-CY5.5EA 2190 PLN
BOSSBS-2456R-CY5.5
Anti-TNFSF18 Rabbit Polyclonal Antibody (Cy5.5®)
Przeciwciała
Cytokine that binds to TNFRSF18/AITR/GITR. Regulates T-cell responses. Can function as costimulator and lower the threshold for T-cell activation and T-cell proliferation. Important for interactions between activated T-lymphocytes and endothelial cells. Mediates activation of NF-kappa-B.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TNFSF18
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR