Drukuj

Anti-Snake poison Protein Rabbit Polyclonal Antibody (Cy7®)

Dostawca: Bioss
BOSSBS-0763R-CY7EA 4310 PLN
BOSSBS-0763R-CY7
Anti-Snake poison Protein Rabbit Polyclonal Antibody (Cy7®)
Przeciwciała
Snake poison Protein

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Snake poison Protein
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR